Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc standard solution, for AAS, 1 mg/ml Zn in 2-5% HCl

Catalog No. ac196465000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196465000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC196465000 Supplier Thermo Scientific Chemicals Supplier No. 196465000
Only null left
Add to Cart

CAS: 7440-66-6 | Zn | 65.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc standard solution
Name Note 1 mg/ml Zn in 2% HCl
Packaging Plastic bottle
CAS 7440-66-6, 7647-01-0, 7732-18-5, 7646-85-7
Density 1.01g/mL
Physical Form Liquid
Molecular Formula Zn
Concentration 997 to 1003 μg/mL of Zn (2+) in 2% HCl (20°C)
MDL Number MFCD00011291
Color Colorless
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES [Zn]
IUPAC Name zinc
Molecular Weight (g/mol) 65.38
Quantity 500 mL
PubChem CID 23994
ChEBI CHEBI:30185
Formula Weight 65.38
Grade AAS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.