Learn More
Hach Company Zinc Standard Solution, 1000 mg/L as Zn (NIST), 100 mL
Catalog No. 501994945 Shop All Hach Company Products
Encompass
For accuracy checks in zinc determinations. (HCl matrix)
This item is not returnable. View return policy
Catalog No. 50-199-4945 Supplier Hach Company Supplier No. 1417742
For accuracy checks in zinc determinations. (HCl matrix)