Learn More
Medchemexpress LLC Zinc stearate 100g
SDP
Catalog No. 5000205872
Encompass_Preferred
Zinc stearate is a Zinc distearate Zinc stearate can be used as an excipient such as lubricants for tablets and capsules Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1][2]
Catalog No. 50-002-05872 Supplier Medchemexpress LLC Supplier No. HYW099569100G
Zinc stearate is a Zinc distearate Zinc stearate can be used as an excipient such as lubricants for tablets and capsules Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1][2]