Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Zinc, Strips, 2 in. x 1/4 in.
SCI_ED

Catalog No. S25910 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 Ea.
Packaging:
Strips (2 in. × 1/4 in.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25910 10 Ea. Strips (2 in. × 1/4 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25910
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 7440-20-0 , 76093-98-6
Synonym 1 mmHg at 487degreeC

Specifications

Quantity 10 Ea.
Ignition Point 460°C
Odor Odorless
Packaging Strips (2 in. × 1/4 in.)
Vapor Pressure 1 mmHg at 487°C
Form Solid
Assay Percent Range 100% w/w
Linear Formula Zn
Chemical Name or Material Zinc, Strips, 2 in. × 1/4 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.