Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Sulfate, 0.0200 M (M/50), Ricca Chemical
SDP

Catalog No. R96570001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R96570001A 1 L
R9657000500 500 mL
R96570004A 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R96570001A Supplier Ricca Chemical Company Supplier No. R96570001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
IUPAC Name zinc(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 L
Grade Laboratory
Packaging Natural Poly Bottle
Chemical Name or Material Zinc Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.