Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Sulfate, 10% (w/v), Ricca Chemical
SDP

Catalog No. R962000020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R962000020F 20 L Cubitainer
9620-16 500 mL Natural Poly Bottle
962032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R962000020F Supplier Ricca Chemical Company Supplier No. R962000020F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

APHA/ASTM/EPA Testing Reagent

Specifications

Color Clear colorless
Concentration 10% (w/v)
pH Range ∼ 4.5
Volume (Metric) 20 L
Format Liquid
Packaging Cubitainer
Quantity 20 L
Type Zinc Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.