Learn More
Biotang Inc ZINC SULFATE 100 mM 50 ML, MT-280, 100 mM Zinc Sulfate solution (ZnSO4). Prepared in 18.2 megohms water and filtered through 0.22 micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50152276 Shop All BIOTANG Inc Products
Encompass_Preferred
ZINC SULFATE 100 mM 50 ML, MT-280, 100 mM Zinc Sulfate solution (ZnSO4). Prepared in 18.2 megohms water and filtered through 0.22 micron filter. Shipped at room temperature. Store at room temperature..
Catalog No. 50-152-276 Supplier Biotang Inc Supplier No. BTBB823
ZINC SULFATE 100 mM 50 ML, MT-280, 100 mM Zinc Sulfate solution (ZnSO4). Prepared in 18.2 megohms water and filtered through 0.22 micron filter. Shipped at room temperature. Store at room temperature..