Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc sulfate heptahydrate | 7446-20-0 | 1 ML
SDP

Catalog No. 5000421739
Encompass_Preferred

Zinc sulfate heptahydrate is the hydrated form of zinc sulfate, recognized for its use as a dietary supplement to address zinc deficiency and aid in prevention for high-risk individuals. This product is provided for research applications.

  • Used as a dietary supplement for zinc deficiency
  • Intended for research use only
  • High purity (99.8%)
  • Molecular weight of 287.55
  • Molecular formula H14O11SZn
  • Appears as a white to off-white solid

Catalog No. 50-004-21739 Supplier Medchemexpress LLC Supplier No. HYN302510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinc sulfate heptahydrate is the hydrated form of zinc sulfate, recognized for its use as a dietary supplement to address zinc deficiency and aid in prevention for high-risk individuals. This product is provided for research applications.

  • Used as a dietary supplement for zinc deficiency
  • Intended for research use only
  • High purity (99.8%)
  • Molecular weight of 287.55
  • Molecular formula H14O11SZn
  • Appears as a white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.