Learn More
Specifications
Specifications
| Melting Point | 100°C |
| Color | White |
| CAS Max % | 100 |
| Physical Form | Crystals |
| Quantity | 500 g |
| Linear Formula | Zn(NO3)2·6H2O |
| Solubility Information | Soluble in water |
| Formula Weight | 297.48 |
| Percent Purity | 99.0 to 103.0% |
| Grade | ACS |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Very toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Wash exposed skin thoroughly after handling. Do not eat, drink or smoke when using this product. Avoid release to the environment. If swallowed: Rinse mouth. Call a poison center/doctor if you feel unwell. Collect spillage. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.