Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Sulfate Heptahydrate, MACCO

Catalog No. 502985607
Encompass
Catalog No. 50-298-5607 Supplier Thermo Scientific Production Chemicals and Services Supplier No. M53004070120

Please call customer service for current pricing. 1-800-766-7000

Chemical Identifiers

CAS 7446-20-0
Molecular Weight (g/mol) 287.6 g/mol

Specifications

Physical Form Solid
Quantity 12 kg poly drum Curtec + 2 x PE liner inside
Percent Purity 99 to 100%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.