Learn More
Strem, An Ascensus Company CAS# 7446-19-7. 250g. Zinc sulfate monohydrate, 99%. MFCD00149893
SDP
Catalog No. 5090115001
Encompass
CAS# 7446-19-7. 250g. Zinc sulfate monohydrate, 99%. MFCD00149893. Molecular Weight: 161.43 (179.45). Molecular Formula: ZnSO4∙H2O. Color/form: white pwdr. Strem# 93-3045. http://www.strem.com/catalog/v/93-3045/
Catalog No. 50-901-15001 Supplier Strem, An Ascensus Company Supplier No. 933045250G
CAS# 7446-19-7. 250g. Zinc sulfate monohydrate, 99%. MFCD00149893. Molecular Weight: 161.43 (179.45). Molecular Formula: ZnSO4∙H2O. Color/form: white pwdr. Strem# 93-3045. http://www.strem.com/catalog/v/93-3045/