Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Sulfate, Monohydrate, FCC, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18607887 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-887 50 kg Poly Bottle
18-607-884 500 g Poly Bottle
18-607-886 12 kg Poly Bottle
18-607-885 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-887 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z110050KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z1100, 7446-19-7, ZnSO4·H2O

Spectrum™ Chemical Zinc Sulfate, Monohydrate, FCC is used as a nutritional supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7446-19-7
Molecular Formula H2O5SZn
Molecular Weight (g/mol) 179.45
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
IUPAC Name zinc(2+) hydrate sulfate
SMILES O.[Zn++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 98.0 to 100.5%
Grade FCC
Packaging Poly Bottle
Chemical Name or Material Zinc Sulfate, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.