Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc sulfate monohydrate, Zn 35.5%

Catalog No. p-7049365
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA14160A1 1 kg
AA14160A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA14160A1 Supplier Thermo Scientific Chemicals Supplier No. 014160A1
Only null left
Add to Cart

CAS: 7446-19-7 | H2O5SZn | 179.451 g/mol

Zinc sulfate monohydrate is used in calico-printing, preserving wood and skins, electrolytes for zinc plating, bleaching paper and clarifying glue. It is also used as a chemical reagent, coagulant in the preparation of rayon, mordant in dyeing and as a zinc source in animal feeds. In medicine, it is used as an astringent and emetic. Further, it is a precursor to the pigment lithopone. In addition to this, it is used to supply zinc in fertilizers, agricultural sprays, in electrolytes for zinc plating and as a mordant in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-19-7
Molecular Formula H2O5SZn
Molecular Weight (g/mol) 179.451
MDL Number MFCD00149893
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
Synonym zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
PubChem CID 62639
IUPAC Name zinc;sulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[Zn+2]

Specifications

Physical Form Coarse Powder
Quantity 1 kg
UN Number UN3077
Merck Index 14,10159
Solubility Information Soluble in water and alcohol.
Formula Weight 179.45 (161.43 Anhydrous)
Concentration or Composition (by Analyte or Components) Zn 35.5%
Chemical Name or Material Zinc sulfate monohydrate
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 231-793-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.