Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc Sulfide

Catalog No. 33327150G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-327-150G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-327-150G Supplier Sigma Aldrich Supplier No. 33327150G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Uses:Preparation of flexible transparent conductive coatings essential for fabrication of a variety of printed electronic devices such as flexible displays and solar cellsPrepare a composite CdS-ZnS/Zirconium-titanium phosphate (ZTP) photocatalyst for hydrogen production under visible lightPrepare light-controlled bioelectrochemical sensors based on CdSe/ZnS quantum dotsCatalyst for photocatalytic degradation of organic pollutantsPreparation of color tunable light-emitting diodes (LEDs)Prepare (CdS-ZnS)-TiO2 combined photocatalysts for electricity production via photoelectrocatalysisCatalyst for synthesis of spirooxindole derivatives in aqueous medium via Knoevenagel condensation followed by Michael additionPrepare CdSe/ZnS q uantum dots for chemiluminescent and chemiluminescence resonance energy transfer (CRET) detection of DNA, metal ions, and aptamer-substrate complexesPreparation of ZnS nanocrystals for ultrasensitive protein detection in terms of multiphonon resonance Raman scattering

Specifications

CAS 10428-19-0
Melting Point 109°C to 112°C
Molecular Formula C16H36Cl2OSn2
Linear Formula {[CH3(CH2)3]2Sn(Cl)}2O
MDL Number MFCD00074921
Quantity 10 g
Synonym Zinc sulphide
Molecular Weight (g/mol) 552.78 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.