Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc Sulfide

Catalog No. 244627100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-462-7100G 100 g
24-462-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-462-7100G Supplier Sigma Aldrich Supplier No. 244627100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zinc sulfide can be used:To prepare flexible transparent conductive coatings essential for fabrication of a variety of printed electronic devices such as flexible displays and solar cells.To prepare a composite CdS-ZnS/zirconium-titanium phosphate (ZTP) photocatalyst for hydrogen production under visible light.To prepare light-controlled bioelectrochemical sensors based on CdSe/ZnS quantum dots.As a catalyst for the photocatalytic degradation of organic pollutants.In the preparation of color tunable light-emitting diodes (LEDs).To prepare (CdS-ZnS)-TiO2 combined photocatalysts for electricity production via photoelectrocatalysis.As a catalyst for the synthesis of spirooxindole derivatives in aqueous medium via Knoevenagel condensation followed by Michael addition.To prepare CdSe/ZnS quantum dots for chemiluminescent and chemiluminescence resonance energy transfer (CRET) detection of DNA, metal ions, and aptamer-substrate complexes.To prepare ZnS nanocrystals for ultrasensitive protein detection in terms of multiphonon resonance Raman scattering.

Specifications

CAS 1314-98-3
Quantity 100 g
Molecular Formula SZn
Linear Formula ZnS
MDL Number MFCD00011301
Synonym Zinc sulphide
Molecular Weight (g/mol) 97.46 g/mol
Density 4.1 g/mL at 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.