Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc sulfide, 99.995% (metals basis)

Catalog No. p-4408060
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4545409 10 g
AA4545418 50 g
AA4545430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4545409 Supplier Thermo Scientific Chemicals Supplier No. 04545409
Only null left
Add to Cart

CAS: 1314-98-3 | SZn | 97.44 g/mol

Zinc sulfide is used in preparation of flexible transparent conductive coatings essential for fabrication of a variety of printed electronic devices such as flexible displays and solar cells. Used to prepare a composite CdS-ZnS/Zirconium-titanium phosphate (ZTP) photocatalyst for hydrogen production under visible light and also applied for light-controlled bioelectrochemical sensors based on CdSe/ZnS quantum dots. It is used for preparation of color tunable light-emitting diodes (LEDs) and also applied to prepare (CdS-ZnS)-TiO2 combined photocatalysts for electricity production via photoelectrocatalysis. It is used as a catalyst for synthesis of spirooxindole derivatives in aqueous medium via Knoevenagel condensation followed by Michael addition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-98-3
Molecular Formula SZn
Molecular Weight (g/mol) 97.44
MDL Number MFCD00011301
InChI Key DRDVZXDWVBGGMH-UHFFFAOYSA-N
Synonym zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
PubChem CID 14821
IUPAC Name sulfanylidenezinc
SMILES [S--].[Zn++]

Specifications

Melting Point 1700°C
Physical Form Pieces
Quantity 10 g
Assay Percent Range 99.995% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 97.43
Density 4.1
Chemical Name or Material Zinc sulfide
EINECSNumber 215-251-3
RTECSNumber ZH5400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.