Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc sulphate | 7733-02-0 | 99.9% | 25 G
SDP

Catalog No. 5000414085
Encompass_Preferred

Zinc sulphate is a biological molecule intended for research use. It appears as a white to off-white solid or crystalline powder with a purity of 99.9%. It is stable under recommended storage conditions, which include room temperature in a dry and cool environment, or in solvent at -80°C for 1 year or -20°C for 6 months. It has good solubility in water.

  • Biological molecule
  • For research use only
  • Purity of 99.9%
  • Appears as a white to off-white solid or crystalline powder
  • Store at room temperature, dry and cool
  • Store in solvent at -80°C for 1 year; -20°C for 6 months
  • Soluble in H2O at 50 mg/mL

Catalog No. 50-004-14085 Supplier Medchemexpress LLC Supplier No. 25GHYB1820
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinc sulphate is a biological molecule intended for research use. It appears as a white to off-white solid or crystalline powder with a purity of 99.9%. It is stable under recommended storage conditions, which include room temperature in a dry and cool environment, or in solvent at -80°C for 1 year or -20°C for 6 months. It has good solubility in water.

  • Biological molecule
  • For research use only
  • Purity of 99.9%
  • Appears as a white to off-white solid or crystalline powder
  • Store at room temperature, dry and cool
  • Store in solvent at -80°C for 1 year; -20°C for 6 months
  • Soluble in H2O at 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.