Learn More
Strem, An Ascensus Company CAS# 13826-88-5. 25g. Zinc tetrafluoroborate hydrate, 98%. MFCD00150371
SDP
Supplier:  Strem, An Ascensus Company 93301025G
Encompass
CAS# 13826-88-5. 25g. Zinc tetrafluoroborate hydrate, 98%. MFCD00150371. Molecular Weight: 238.98. Molecular Formula: Zn(BF4)2∙XH2O. Color/form: white xtl. Strem# 93-3010. http://www.strem.com/catalog/v/93-3010/
Catalog No. 50-901-14959