Learn More
Perkin Elmer US LLC Zinc (Zn) Pure Plus Standard, 1 µg/mL, 2% HNO3, 125 mL
Catalog No. 502111865
Encompass_Preferred
Each Single Element Calibration Standard for ICP-MS is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
Catalog No. 50-211-1865 Supplier Perkin Elmer US LLC Supplier No. N9304289
Each Single Element Calibration Standard for ICP-MS is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.