Learn More
Cayman Chemical ZINC00881524, Purity: ≥98%, CAS: 557782-81-7, 100 mg
SDP
Catalog No. 502988854
Encompass_Preferred
A ROCK inhibitor; decreases levels of ROCK1 in T47D and CAMA-1 breast cancer cells; decreases proliferation of T47D and CAMA-1 cells when used in combination with CNN1 knockdown
Catalog No. 50-298-8854 Supplier Cayman Chemical Supplier No. 30788100 MG
A ROCK inhibitor; decreases levels of ROCK1 in T47D and CAMA-1 breast cancer cells; decreases proliferation of T47D and CAMA-1 cells when used in combination with CNN1 knockdown