Learn More
Medchemexpress LLC Zinc05007751 | 591239-68-8 | 99.3% | 50 MG
SDP
Catalog No. 5000231071
Encompass_Preferred
ZINC05007751 is a research-grade small-molecule inhibitor of NIMA-related kinase 6 (NEK6) with a reported IC50 of approximately 3.4 μM. It exhibits antiproliferative activity across multiple human cancer cell lines and has reported synergistic effects with cisplatin and paclitaxel in BRCA2-mutated ovarian cancer models. The compound has formula C18H12N2O3, molecular weight 304.30 g/mol, and listed purity 99.33%.
  • Inhibits NEK6 with an IC50 of ~3.4 μM.
  • Shows antiproliferative activity in various human cancer cell lines.
  • Reported synergy with cisplatin and paclitaxel in BRCA2-mutated ovarian cancer models.
  • High purity, suitable for research applications.
  • Supplied as a 50 mg research pack; soluble in DMSO.
Catalog No. 50-002-31071 Supplier Medchemexpress LLC Supplier No. HY12263950MG
ZINC05007751 is a research-grade small-molecule inhibitor of NIMA-related kinase 6 (NEK6) with a reported IC50 of approximately 3.4 μM. It exhibits antiproliferative activity across multiple human cancer cell lines and has reported synergistic effects with cisplatin and paclitaxel in BRCA2-mutated ovarian cancer models. The compound has formula C18H12N2O3, molecular weight 304.30 g/mol, and listed purity 99.33%.
  • Inhibits NEK6 with an IC50 of ~3.4 μM.
  • Shows antiproliferative activity in various human cancer cell lines.
  • Reported synergy with cisplatin and paclitaxel in BRCA2-mutated ovarian cancer models.
  • High purity, suitable for research applications.
  • Supplied as a 50 mg research pack; soluble in DMSO.