Learn More
TARGETMOL CHEMICALS INC ZINC12409120 5MG
SDP
Catalog No. 502579546
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ZINC12409120 (R-4584) is a selective ERK inhibitor.ZINC12409120 inhibits ERK with an IC50 of 5.0 uM.ZINC12409120 inhibits ERK activity by interfering with FGF23(alpha)-Klotho interaction. purity: 99%
Catalog No. 50-257-9546 Supplier TARGETMOL CHEMICALS INC Supplier No. T611225MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ZINC12409120 (R-4584) is a selective ERK inhibitor.ZINC12409120 inhibits ERK with an IC50 of 5.0 uM.ZINC12409120 inhibits ERK activity by interfering with FGF23(alpha)-Klotho interaction. purity: 99%