Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zincon Sodium Salt, MP Biomedicals™

Catalog No. ICN15831705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15831705 5 g
ICN15831701 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15831705 Supplier MP Biomedicals, Inc Supplier No. 0215831705
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62625-22-3
Molecular Formula C20H16N4NaO6S
Molecular Weight (g/mol) 463.42
InChI Key IABRINWJUBAIIE-JJECXDOKSA-N
Synonym 2-Carboxy-2'-hydroxy-5'-sulfoformazylbenzene
PubChem CID 131856391
IUPAC Name 2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium
SMILES C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na]

Specifications

Quantity 5 g
Formula Weight 462.4
Physical Form Solid
Chemical Name or Material Zincon Sodium Salt
EINECSNumber 263-651-1
Recommended Storage Store at 2° to 8°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.