Learn More
Hach Company ZincoVer 5 Zinc Reagent Powder Pillows, 20 mL, pk/100
Supplier:  Hach Company 2106669
Encompass
For determination of zinc by the ZincoVer 5 zincon method. USEPA approved for wastewater analysis - Hach method 8009. Range: 0.01 to 3.00 mg/L Zn. Sample size: 20-mL. Pack of 100 Powder Pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-5410