Learn More
Hach Company ZincoVer 5 Zinc Reagent Powder Pillows, 50 mL, pk/25
Supplier:  Hach Company 1403268
Encompass
For zinc determination by the zincon method. EPA compliant - Hach method 8009. Sample size: 50 mL. Pack of 25 powder pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-4890