Learn More
Grainger ZING Lockout Kit, Electrical Valve
Catalog No. NC2229920
Encompass_Preferred
The Zing Lockout Kit Electrical Valve is a lockout kit designed to lock out electrical valves. It includes a lockout hasp, valve lockout, and a danger tag. The hasp accommodates up to 6 locks and can be used to lockout breakers, switches, valves, and other energy sources. The valve lockout can lock ball valves in the off position. The danger tag can be used for labeling and identification. This kit provides a way to lock out energy sources during maintenance and repairs to prevent accidental startup and ensure safety. The components are made of durable polypropylene and stainless steel. It meets OSHA lockout/tagout requirements.
This item is not returnable. View return policy
Catalog No. NC2229920 Supplier Grainger Supplier No. 28AV86
The Zing Lockout Kit Electrical Valve is a lockout kit designed to lock out electrical valves. It includes a lockout hasp, valve lockout, and a danger tag. The hasp accommodates up to 6 locks and can be used to lockout breakers, switches, valves, and other energy sources. The valve lockout can lock ball valves in the off position. The danger tag can be used for labeling and identification. This kit provides a way to lock out energy sources during maintenance and repairs to prevent accidental startup and ensure safety. The components are made of durable polypropylene and stainless steel. It meets OSHA lockout/tagout requirements.