Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zingerone, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012802 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2802 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2802 Supplier MilliporeSigma Supelco Supplier No. 8878750MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 122-48-5
Density 1.14 g/mL (at 25°C)
Linear Formula 4-(HO)C6H3-3-(OCH3)CH2 CH2 COCH3
MDL Number MFCD00048232
Melting Point 40°C to 41°C
Boiling Point 141°C (0.5 mmHg)
Synonym Vanillylacetone; Zingerone
Type Analytical Standard
Quantity 50 mg
Formula Weight 194.23
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.