Learn More
Medchemexpress LLC Zingiberen newsaponin | 91653-50-8 | 99.7% | 1047.18 g/mol | C51H82O22 | 10 MG
SDP
Catalog No. 5000289368
Encompass_Preferred
Zingiberen newsaponin is an orally active steroid saponin used in preclinical research with reported anticancer and neuroprotective activities. It is provided as a solid for laboratory studies and is intended for research use only.
  • Orally active steroid saponin with reported in vitro and in vivo activity.
  • Exerts anti-hepatocellular carcinoma effects by inhibiting autophagy and the AKR1C1/JAK2/STAT3 pathway.
  • Activates oxidative stress and mitochondrial pathways, promoting cancer cell apoptosis.
  • Alleviates cerebral ischemia-reperfusion injury and reduces pro-inflammatory cytokines.
  • Enhances antioxidant enzyme activity and protects nerve cells.
  • Solid form; white to off-white appearance; store sealed away from moisture and light.
  • Analytical purity reported at 99.68%.
Catalog No. 50-002-89368 Supplier Medchemexpress LLC Supplier No. HYN228210MG
Zingiberen newsaponin is an orally active steroid saponin used in preclinical research with reported anticancer and neuroprotective activities. It is provided as a solid for laboratory studies and is intended for research use only.
  • Orally active steroid saponin with reported in vitro and in vivo activity.
  • Exerts anti-hepatocellular carcinoma effects by inhibiting autophagy and the AKR1C1/JAK2/STAT3 pathway.
  • Activates oxidative stress and mitochondrial pathways, promoting cancer cell apoptosis.
  • Alleviates cerebral ischemia-reperfusion injury and reduces pro-inflammatory cytokines.
  • Enhances antioxidant enzyme activity and protects nerve cells.
  • Solid form; white to off-white appearance; store sealed away from moisture and light.
  • Analytical purity reported at 99.68%.