Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zingibroside R1 | 80930-74-1 | 5 MG
SDP

Catalog No. 5000419518
Encompass_Preferred

Zingibroside R1 is a dammaranae-type triterpenoid saponin isolated from the rhizomes, taproots, and lateral roots of Panax japonicus C. A. Meyer. It exhibits anti-tumor effects, anti-angiogenic activity, and anti-HIV-1 activity. It also shows inhibitory effects on 2-deoxy-D-glucose (2-DG) uptake by EAT cells with an IC50 of 91.3 μM. This compound has a molecular weight of 794.97, a chemical formula of C42H66O14, and appears as a white to off-white solid.

  • Isolated from Panax japonicus C. A. Meyer
  • Exhibits anti-tumor effects
  • Shows anti-angiogenic activity
  • Possesses anti-HIV-1 activity
  • Inhibits 2-deoxy-D-glucose (2-DG) uptake by EAT cells (IC50=91.3 μM)
  • Molecular weight: 794.97
  • Chemical formula: C42H66O14
  • Appearance: white to off-white solid
  • Purity: 99.47%

Catalog No. 50-004-19518 Supplier Medchemexpress LLC Supplier No. HYN69245MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zingibroside R1 is a dammaranae-type triterpenoid saponin isolated from the rhizomes, taproots, and lateral roots of Panax japonicus C. A. Meyer. It exhibits anti-tumor effects, anti-angiogenic activity, and anti-HIV-1 activity. It also shows inhibitory effects on 2-deoxy-D-glucose (2-DG) uptake by EAT cells with an IC50 of 91.3 μM. This compound has a molecular weight of 794.97, a chemical formula of C42H66O14, and appears as a white to off-white solid.

  • Isolated from Panax japonicus C. A. Meyer
  • Exhibits anti-tumor effects
  • Shows anti-angiogenic activity
  • Possesses anti-HIV-1 activity
  • Inhibits 2-deoxy-D-glucose (2-DG) uptake by EAT cells (IC50=91.3 μM)
  • Molecular weight: 794.97
  • Chemical formula: C42H66O14
  • Appearance: white to off-white solid
  • Purity: 99.47%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.