Learn More
Medchemexpress LLC Zingibroside R1 | 80930-74-1 | MFCD32201173 | 10 MG
SDP
Catalog No. 5000419347
Encompass_Preferred
Zingibroside R1 is a dammaranae-type triterpenoid saponin isolated from the rhizomes, taproots, and lateral roots of Panax japonicas C. A. Meyer. It exhibits anti-tumor and anti-angiogenic activity.
  • Exhibits anti-tumor effects
  • Shows anti-angiogenic activity
  • Possesses anti-HIV-1 activity
  • Inhibits 2-deoxy-D-glucose (2-DG) uptake by EAT cells (IC50=91.3 μM)
Catalog No. 50-004-19347 Supplier Medchemexpress LLC Supplier No. HYN692410MG
Zingibroside R1 is a dammaranae-type triterpenoid saponin isolated from the rhizomes, taproots, and lateral roots of Panax japonicas C. A. Meyer. It exhibits anti-tumor and anti-angiogenic activity.
  • Exhibits anti-tumor effects
  • Shows anti-angiogenic activity
  • Possesses anti-HIV-1 activity
  • Inhibits 2-deoxy-D-glucose (2-DG) uptake by EAT cells (IC50=91.3 μM)