Learn More
AdipoGen Zinpyr-1, 288574-78-7, MFCD04039712, 20 mg
SDP
Supplier:  AdipoGen CDXZ0001M020
Encompass_Preferred
MF: C46H36Cl2N6O5, MW: 823.72, ≥98%, Appearance: Salmon pink powder. Membrane permeable fluorescein-based probe suitable for the fluorometric detection of Zn2+. In living cells Zinpyr-1 mainly stains the Golgi or a Golgi-associated vesicle. It undergoes a shift in excitation from 515 to 507nm with increasing zinc concentrations using an emission filter over 513-558nm. A stock solution of 1mM (823ug/ml) should be prepared to stain brain sections. This stock solution can be further diluted in 0.9% saline to get a working solution with a final concentration of 10-20uM.
Catalog No. 50-596-113