Learn More
AdipoGen Zinpyr-1
SDP
Supplier:  AdipoGen CDXZ0001M125
Encompass_Preferred
Chemical. CAS 288574-78-7. Formula C46H36Cl2N6O5. MW 823.72. Synthetic. Membrane permeable fluorophore suitable for the fluorometric detection of Zn2. In living cells Zinpyr-1 mainly stains the Golgi or a Golgi-associated vesicle.
Catalog No. 50-255-9892