Learn More
Cayman Chemical ZInquIn ethyl ester 10mg
SDP
Catalog No. 502972760
Encompass_Preferred
A cell-permeable, quinolone-based fluorescent probe used as a zinc indicator (excitation 368 nm, emission 490 nm); used to monitor intracellular zinc fluxes associated with apoptosis
Catalog No. 50-297-2760 Supplier Cayman Chemical Supplier No. 1513310 MG
A cell-permeable, quinolone-based fluorescent probe used as a zinc indicator (excitation 368 nm, emission 490 nm); used to monitor intracellular zinc fluxes associated with apoptosis