Learn More
AdipoGen Zinquin ethyl ester
SDP
Supplier:  AdipoGen CDXZ0013M050
Encompass_Preferred
Chemical. CAS 181530-09-6. Formula C21H22N2O5S. MW 414.47. Synthetic. ZincII specific fluorophores probes.
Catalog No. 50-255-9871