Learn More
Cayman Chemical Zinquin ethyl ester, Purity: ≥98%, CAS: 181530-09-6, 5 mg
SDP
Catalog No. 502971756
Encompass_Preferred
A cell-permeable, quinolone-based fluorescent probe used as a zinc indicator (excitation 368 nm, emission 490 nm); used to monitor intracellular zinc fluxes associated with apoptosis
Catalog No. 50-297-1756 Supplier Cayman Chemical Supplier No. 151335 MG
A cell-permeable, quinolone-based fluorescent probe used as a zinc indicator (excitation 368 nm, emission 490 nm); used to monitor intracellular zinc fluxes associated with apoptosis