Learn More
Abcam Zinquin ethyl ester, Zn2+ selective fluorescent probe, 50MG
Catalog No. 502705167
Encompass_Preferred
MW 414.5 Da, Purity >99%. Cell-permeable Zn2+ selective fluorescent probe. Zinquin (ab145451) precursor. Labels micromolar concentrations of intracellular Zn2+ ions with an intense extranuclear fluorescence emitting in the blue range.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5167 Supplier Abcam Supplier No. AB14547850MG
MW 414.5 Da, Purity >99%. Cell-permeable Zn2+ selective fluorescent probe. Zinquin (ab145451) precursor. Labels micromolar concentrations of intracellular Zn2+ ions with an intense extranuclear fluorescence emitting in the blue range.
The product is subject to the following: Abcam Restricted Use Statement