Learn More
AdipoGen Zinquin free acid
SDP
Catalog No. 502559870
Encompass_Preferred
Chemical. CAS 151606-29-0. Formula C19H18N2O5S. MW 386.42. Synthetic. Used to detect zinc in cells by UV fluorescence.
Catalog No. 50-255-9870 Supplier AdipoGen Supplier No. CDXZ0014M001
Add to Cart
Add to Cart
Chemical. CAS 151606-29-0. Formula C19H18N2O5S. MW 386.42. Synthetic. Used to detect zinc in cells by UV fluorescence.