Learn More
Biotium Zinquin Free Acid
Catalog No. 501964710
Encompass_Preferred
Zinquin is an UV-excitable, blue fluorescent zinc indicator. Zinc is believed to be involved in the suppression of apoptosis and thought to play important roles in many neural activities. Ex/Em (with Zn2)= 364/385 nm; Off-white solid soluble in DMSO; Store at 4°C and protect from light; C19H18N2O5S; MW: 386.42.
Catalog No. 50-196-4710 Supplier Biotium Supplier No. 52022
Zinquin is an UV-excitable, blue fluorescent zinc indicator. Zinc is believed to be involved in the suppression of apoptosis and thought to play important roles in many neural activities. Ex/Em (with Zn2)= 364/385 nm; Off-white solid soluble in DMSO; Store at 4°C and protect from light; C19H18N2O5S; MW: 386.42.