Learn More
Abcam Zinquin, Zn2+ selective fluorescent probe, 25MG
Catalog No. 502705181
Encompass_Preferred
MW 386.4 Da, Purity >95%. Intracellular Zn2+ selective fluorescent probe. Forms zinquin-Zn2+ complexes in either 1:1 or 2:1 ratios (Kd values are 370 and 850 nM respectively). Zinquin ethyl ester (ab145478) analog. Labels micromolar concentrations of intracellular Zn2+ ions with an intense extranuclear fluorescence emitting in the blue range.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5181 Supplier Abcam Supplier No. AB14545125MG
MW 386.4 Da, Purity >95%. Intracellular Zn2+ selective fluorescent probe. Forms zinquin-Zn2+ complexes in either 1:1 or 2:1 ratios (Kd values are 370 and 850 nM respectively). Zinquin ethyl ester (ab145478) analog. Labels micromolar concentrations of intracellular Zn2+ ions with an intense extranuclear fluorescence emitting in the blue range.
The product is subject to the following: Abcam Restricted Use Statement