Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinquin

Catalog No. Z23765MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z23765MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Z23765MG Supplier Sigma Aldrich Supplier No. Z23765MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zinquin has been used to detect intracellular zinc.Zinquin is a membrane-permeant fluorophore. It is used to detect zinc in cells by UV fluorescence. Zinquin ethyl ester can be used to monitor the change in intracellular concentrations of Zn2+ in thymocytes and hepatocytes. The ester is excited at 368nm aFree acid or salt as available

Specifications

CAS 33758-15-5
Density 0.827 g/mL at 25°C
Molecular Formula C10H22O
Linear Formula CH3(CH2)7CH(OH)CH3
MDL Number MFCD03427199
Quantity 1 g
Molecular Weight (g/mol) 158.28 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.