Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinterol hydrochloride | 38241-28-0 | 99.7% | C19H27ClN2O4S | 1 ML
SDP

Catalog No. 5000355518
Encompass_Preferred

Zinterol hydrochloride is a potent and selective β2-adrenoceptor agonist. It increases ICa in a concentration-dependent manner with an EC50 of 2.2 nM. This compound has been shown to induce ventricular arrhythmias in conscious heart failure rabbits at specific dosages. It is intended strictly for research purposes.

  • Potent and selective β2-adrenoceptor agonist.
  • Increases ICa with an EC50 of 2.2 nM.
  • Induces ventricular arrhythmias in heart failure animal models.
  • For research use only.
  • High purity of 99.7%.
  • Solid form can be stored at 4°C, sealed, away from moisture.
  • Solution form can be stored at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture.

Catalog No. 50-003-55518 Supplier Medchemexpress LLC Supplier No. HY14304A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinterol hydrochloride is a potent and selective β2-adrenoceptor agonist. It increases ICa in a concentration-dependent manner with an EC50 of 2.2 nM. This compound has been shown to induce ventricular arrhythmias in conscious heart failure rabbits at specific dosages. It is intended strictly for research purposes.

  • Potent and selective β2-adrenoceptor agonist.
  • Increases ICa with an EC50 of 2.2 nM.
  • Induces ventricular arrhythmias in heart failure animal models.
  • For research use only.
  • High purity of 99.7%.
  • Solid form can be stored at 4°C, sealed, away from moisture.
  • Solution form can be stored at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.