Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinterol hydrochloride | 38241-28-0 | MFCD00867031 | 99.5% | 414.95 g/mol | C19H27ClN2O4S | 5 MG
SDP

Catalog No. 5000221227
Encompass_Preferred

Zinterol hydrochloride is the hydrochloride salt of a potent, selective β2-adrenoceptor agonist supplied for research use. It is provided as a high-purity solid and in DMSO solution formats for in vitro pharmacology studies involving β2 receptor activation and modulation of calcium currents.

  • Potent, selective β2-adrenoceptor agonist.
  • Supplied as the hydrochloride salt suitable for research use.
  • High purity (≈99.5%) for reliable experimental results.
  • Molecular weight 414.95 g/mol; formula C19H27ClN2O4S.
  • Soluble in DMSO; 10 mM solution formats available.
  • Pack sizes available including small-scale quantities for screening and assay development.

Catalog No. 50-002-21227 Supplier Medchemexpress LLC Supplier No. HY14304A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinterol hydrochloride is the hydrochloride salt of a potent, selective β2-adrenoceptor agonist supplied for research use. It is provided as a high-purity solid and in DMSO solution formats for in vitro pharmacology studies involving β2 receptor activation and modulation of calcium currents.

  • Potent, selective β2-adrenoceptor agonist.
  • Supplied as the hydrochloride salt suitable for research use.
  • High purity (≈99.5%) for reliable experimental results.
  • Molecular weight 414.95 g/mol; formula C19H27ClN2O4S.
  • Soluble in DMSO; 10 mM solution formats available.
  • Pack sizes available including small-scale quantities for screening and assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.