Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinterol hydrochloride | 38241-28-0 | MFCD00867031 | 99.5% | C19H27ClN2O4S | 10MG
SDP

Supplier:  Medchemexpress LLC HY14304A10MG

Encompass_Preferred

Zinterol hydrochloride is a research-grade β2-adrenoceptor agonist supplied as the hydrochloride salt for pharmacological and electrophysiological studies. It is provided in small research pack sizes and is available as a DMSO solution for ready use in assays and receptor characterization.

  • Potent and selective β2-adrenoceptor agonist.
  • High purity (≈99.5%).
  • Molecular formula C19H27ClN2O4S and molecular weight 414.95.
  • Soluble in DMSO; available as a ready 10 mM solution.
  • Suitable for receptor pharmacology and electrophysiology applications.

Catalog No. 50-002-61502


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.