Learn More
Shields Dina Corporation ZIP 6 X9 X4MIL1000/CS
Catalog No. 50011454
Encompass_Preferred
ZIP RECLOSABLE WRITE-ON BAG, 6in X9in X4MIL, 1000/CS
Catalog No. 50-011-454 Supplier Shields Dina Corporation Supplier No. S498
ZIP RECLOSABLE WRITE-ON BAG, 6in X9in X4MIL, 1000/CS