Learn More
Thomas Scientific 9X12 PLAIN ZIP BG 4MIL PK100
SDP
Catalog No. NC9217133
Encompass_Preferred
ZIP Bag 4mm, 9 x 12 in. Clear, polypropylene bags, Leak resistant, High chemical compatibility, proprietary zipper closure; For storing samples, req. less space than rigid containers
Catalog No. NC9217133 Supplier Thomas Scientific Supplier No. 1304L84
ZIP Bag 4mm, 9 x 12 in. Clear, polypropylene bags, Leak resistant, High chemical compatibility, proprietary zipper closure; For storing samples, req. less space than rigid containers