Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab Zip Closure Specimen Bags

Catalog No. 22505020
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-505-020 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-505-020 Supplier Marketlab Supplier No. 108167YL
Add to Cart
Add to Cart

Easy-open colored design with secure zip closure. Supports specimen organization and transport.

  • Top-Colored Specimen Bags
  • Colored tops can be used for color coding specimens
  • Zipper seal provides fast, easy, and liquid-tight closure
  • Biohazard designation is black symbol on color background
  • Four preprinted check boxes for clear handling instructions
  • Three-wall construction with separate document pouch
  • Bilingual text in English and Spanish
  • 6 W x 9 H in.

Specifications

Autoclavable Not Autoclavable
Closure Type Zip Closure
Color Yellow
Disposable Single-use
Material LDPE
Sterility Non-sterile
Length (English) 9 in.
Width (English) 6 in.
Product Type Specimen Bag
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.