Learn More
Medicus Health Zip Compact Hematocrit Centrifuge
SDP
Catalog No. NC3880463
Encompass
• 12 Place, Zip Compact Hematocrit centrifuge for spinning half-size 40mm capillary tubes
• Attractive unit is very small, conserving valuable counter space in the lab, but also sturdy and heavy with suction-cupped feet for stability
• Digital controls allow speed and time to be adjusted with the touch of a finger and will show real-time speed and time remaining
• Centrifuge is surprisingly quiet, making the lab environment much more peaceful…talking on the phone beside it is not a problem
• Powerful, maintenance-free, brushless motor spins at 12,000 rpm producing PCV results in 3 minutes
• Auto-brake stops the rotor in 10 seconds, and the lid pops open automatically upon completion
• Included EZ Reader card is easy and intuitive to use
• Zip-IQ is the smart choice for any clinic or laboratory
Catalog No. NC3880463 Supplier Medicus Health Supplier No. 4088M1
• 12 Place, Zip Compact Hematocrit centrifuge for spinning half-size 40mm capillary tubes
• Attractive unit is very small, conserving valuable counter space in the lab, but also sturdy and heavy with suction-cupped feet for stability
• Digital controls allow speed and time to be adjusted with the touch of a finger and will show real-time speed and time remaining
• Centrifuge is surprisingly quiet, making the lab environment much more peaceful…talking on the phone beside it is not a problem
• Powerful, maintenance-free, brushless motor spins at 12,000 rpm producing PCV results in 3 minutes
• Auto-brake stops the rotor in 10 seconds, and the lid pops open automatically upon completion
• Included EZ Reader card is easy and intuitive to use
• Zip-IQ is the smart choice for any clinic or laboratory