Learn More
Health Care Logistics Inc ZIP-IT BAG AMBER 6X8IN 100PK
SDP
Catalog No. NC9387282
Encompass
Bag; Zip-It; Amber; 6 x 8; 3mil T; Resistant to U-V light penetration, therefore perfect for storing or dispensing light sensitive drugs; Dimensions: 6L x 8 in. W (152 x 203mm);
Catalog No. NC9387282 Supplier Health Care Logistics Inc Supplier No. 7575
Bag; Zip-It; Amber; 6 x 8; 3mil T; Resistant to U-V light penetration, therefore perfect for storing or dispensing light sensitive drugs; Dimensions: 6L x 8 in. W (152 x 203mm);