Learn More
Cole-Parmer Zip-lip low-density polyethylene bag, 9" x 12"
Supplier:  Cole-Parmer 6010180
Encompass_Preferred
Low-density polyethylene. Zip-lip seal allows dustproof, moisture-proof storage. All bags are single track. Not for use with liquids.
Catalog No. NC0161710