Learn More
ABclonal Technology Zip12 Rabbit pAb
SDP
Supplier:  ABclonal Technology A160061000UL
Encompass_Preferred
Zinc is an essential cofactor for hundreds of enzymes. It is involved in protein, nucleic acid, carbohydrate, and lipid metabolism, as well as in the control of gene transcription, growth, development, and differentiation. SLC39A12 belongs to a subfamily of proteins that show structural characteristics of zinc transporters (Taylor and Nicholson, 2003 [PubMed 12659941]).
Catalog No. 50-001-08209